missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Bis(hydroxymethyl)durene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 256°C |
| Quantity | 1 g |
| Formula Weight | 194.27 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,6-Bis(hydroxymethyl)durene |
Safety and Handling
| TSCA | No |