missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dinitrosalicylic Acid 98.0+%, TCI America™
Supplier: TCI America D0850500G
Specifications
| 3,5-Dinitrosalicylic Acid | |
| 172°C | |
| MFCD00007104 | |
| 2811 | |
| LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
| 228.116 | |
| CHEBI:53648 | |
| ≥98.0% (GC,T) |
| 609-99-4 | |
| C7H4N2O7 | |
| 500 g | |
| 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
| C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] | |
| 11873 | |
| 228.12 | |
| Crystalline Powder |
Chemical Identifiers
| 609-99-4 | |
| 228.116 | |
| LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
| 11873 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] |
| C7H4N2O7 | |
| MFCD00007104 | |
| 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
| CHEBI:53648 |
Safety and Handling
RTECSNumber : VO2965000
TSCA : Yes