accessibility menu, dialog, popup

UserName

OHOHONOONOO

CAS RN 609-99-4

IUPAC Name: 2-hydroxy-3,5-dinitrobenzoic acid
Synonyms: 3,5-dinitrosalicylic acid o-dinitrocarboxylphenol 3,5-dinitrosalicylate o-dncp
Molecular Weight (g/mol): 228.12
Molecular Formula: C7H4N2O7
InChi Key: LWFUFLREGJMOIZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (3)
  • (5)
  • (6)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)