Learn More
3,5-Dinitrobenzyl alcohol, 98%
CAS: 71022-43-0 | C7H6N2O5 | 198.13 g/mol
$162.93 - $1844.74
Chemical Identifiers
| CAS | 71022-43-0 |
|---|---|
| Molecular Formula | C7H6N2O5 |
| Molecular Weight (g/mol) | 198.13 |
| MDL Number | MFCD00007235 |
| InChI Key | GPHYIQCSMDYRGJ-UHFFFAOYSA-N |
| Synonym | 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # |
| PubChem CID | 116978 |
| IUPAC Name | (3,5-dinitrophenyl)methanol |
| SMILES | OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3266906
|
Thermo Scientific Chemicals
H3266906 |
5 g |
Each for $162.93
|
|
|||||
|
AAH3266914
|
Thermo Scientific Chemicals
H3266914 |
25 g |
Each for $540.18
|
|
|||||
|
AAH3266922
|
Thermo Scientific Chemicals
H3266922 |
100 g |
Each for $1,844.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 71022-43-0 | |
| 198.13 | |
| GPHYIQCSMDYRGJ-UHFFFAOYSA-N | |
| 116978 | |
| OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
| C7H6N2O5 | |
| MFCD00007235 | |
| 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # | |
| (3,5-dinitrophenyl)methanol |
Specifications
| 71022-43-0 | |
| C7H6N2O5 | |
| 5 g | |
| 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # | |
| OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O | |
| 198.13 | |
| 198.13 | |
| 3,5-Dinitrobenzyl alcohol |
| 88°C to 91°C | |
| MFCD00007235 | |
| 2054388 | |
| GPHYIQCSMDYRGJ-UHFFFAOYSA-N | |
| (3,5-dinitrophenyl)methanol | |
| 116978 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 275-131-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only