missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzyl alcohol, 98%

CAS: 71022-43-0 | C7H6N2O5 | 198.13 g/mol

Supplier:  Thermo Scientific Chemicals H3266914

 View more versions of this product

Catalog No. AAH3266914

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dinitrobenzyl alcohol
88°C to 91°C
MFCD00007235
2054388
GPHYIQCSMDYRGJ-UHFFFAOYSA-N
(3,5-dinitrophenyl)methanol
116978
98%
71022-43-0
C7H6N2O5
25 g
3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
198.13
198.13

Chemical Identifiers

71022-43-0
198.13
GPHYIQCSMDYRGJ-UHFFFAOYSA-N
116978
OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
C7H6N2O5
MFCD00007235
3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
(3,5-dinitrophenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 275-131-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only