missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzonitrile, 98%

Catalog No. AAA1522314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522314 25 g
AAA1522306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522314 Supplier Thermo Scientific Chemicals Supplier No. A1522314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4110-35-4 | C7H3N3O4 | 193.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4110-35-4
Molecular Formula C7H3N3O4
Molecular Weight (g/mol) 193.12
MDL Number MFCD00007228
InChI Key SSDNULNTQAUNFQ-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-dinitro, 3,5-dinitro-benzonitrile, 3,5-dinitrobenzenecarbonitrile, acmc-209jih, 3,5-bisnitrobenzonitrile, 3,5-dinitrobenzonitrile, wln: wnr cnw ecn, benzonitrile,3,5-dinitro, 1,3-dinitro-5-cyanobenzene, 5-cyano-1,3-dinitrobenzene
PubChem CID 20062
IUPAC Name 3,5-dinitrobenzonitrile
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C#N)[N+]([O-])=O

Specifications

Melting Point 127°C to 130°C
Quantity 25 g
UN Number UN3439
Formula Weight 193.12
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzonitrile
Hazard Category H301-H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 223-889-9
RTECSNumber DI4365000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only