accessibility menu, dialog, popup

UserName

ONONOON

CAS RN 4110-35-4

IUPAC Name: 3,5-dinitrobenzonitrile
Synonyms: benzonitrile, 3,5-dinitro-
Molecular Weight (g/mol): 193.12
Molecular Formula: C7H3N3O4
InChi Key: SSDNULNTQAUNFQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1)C#N)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)