missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylpyrazole 98.0+%, TCI America™
Supplier: TCI America D164725G
Specifications
| 3,5-Dimethylpyrazole | |
| 108°C | |
| 218°C | |
| MFCD00005243 | |
| 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256 | |
| CC1=CC(C)=NN1 | |
| 96.13 | |
| 96.13 | |
| Crystalline Powder |
| 67-51-6 | |
| White-Yellow | |
| C5H8N2 | |
| 25 g | |
| SDXAWLJRERMRKF-UHFFFAOYSA-N | |
| 3,5-dimethyl-1H-pyrazole | |
| 6210 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 67-51-6 | |
| 96.13 | |
| SDXAWLJRERMRKF-UHFFFAOYSA-N | |
| 6210 | |
| CC1=CC(C)=NN1 |
| C5H8N2 | |
| MFCD00005243 | |
| 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256 | |
| 3,5-dimethyl-1H-pyrazole |
Safety and Handling
EINECSNumber : (9)-0605
RTECSNumber : UQ6477600
TSCA : Yes