missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazole 98.0+%, TCI America™

Catalog No. p-7126848
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D164725G 25 g
D1647100G 100 g
D1647500G 500 g
3 options

Catalog No. D164725G

Supplier: TCI America D164725G

Only null left

Chemical Identifiers

CAS 67-51-6
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005243
InChI Key SDXAWLJRERMRKF-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
PubChem CID 6210
IUPAC Name 3,5-dimethyl-1H-pyrazole
SMILES CC1=CC(C)=NN1

Specifications

Melting Point 108°C
Color White-Yellow
Boiling Point 218°C
Quantity 25 g
Formula Weight 96.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylpyrazole
EINECSNumber (9)-0605
RTECSNumber UQ6477600
TSCA Yes