missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylhydrazine hydrochloride, 97%

Catalog No. AAA1133606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1133606 5 g
AAA1133614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1133606 Supplier Thermo Scientific Chemicals Supplier No. A1133606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60481-36-9 | C8H13ClN2 | 172.656 g/mol

3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 60481-36-9
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00052269
InChI Key RSBQANBNDXZFIY-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylhydrazine hydrochloride, 3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenylhydrazinehydrochloride, 3,5-dimethyl phenylhydrazine hydrochloride, 3,5-dimethylphenylhydrazine, hydrazine, 3,5-dimethylphenyl-, monohydrochloride, pubchem6407, ksc491e7r, 1-3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenyl hydrazine-hydrogen chloride 1/1
PubChem CID 12651341
IUPAC Name (3,5-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=CC(=CC(=C1)NN)C.Cl

Specifications

Melting Point 180°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylhydrazine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only