missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$83.89 - $83.89
Specifications
| CAS | 172975-69-8 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C8H11BO2 |
| MDL Number | MFCD00185689 |
| InChI Key | DJGHSJBYKIQHIK-UHFFFAOYSA-N |
Specifications
| 172975-69-8 | |
| C8H11BO2 | |
| DJGHSJBYKIQHIK-UHFFFAOYSA-N | |
| (3,5-dimethylphenyl)boronic acid | |
| 2734349 |
| Crystal-Powder at 20°C | |
| MFCD00185689 | |
| B(C1=CC(=CC(=C1)C)C)(O)O | |
| 149.984 | |
| 149.98 |