missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylacetonitrile, 98%

CAS: 39101-54-7 | C10H11N | 145.205 g/mol

$136.10 - $490.75

Chemical Identifiers

CAS 39101-54-7
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00060304
InChI Key LMUKNQSVBFEUKR-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylacetonitrile, 2-3,5-dimethylphenyl acetonitrile, 3,5-dimethylphenyl acetonitrile, benzeneacetonitrile, 3,5-dimethyl, 3,5-dimethyl-phenyl-acetonitrile, 2-3,5-dimethylphenyl ethanenitrile, acmc-20a3vm, 3.5-dimethylphenylacetonitril, 3,5-dimethylbenzeneacetonitrile, 3,5-dimethylphenylethanenitrile
PubChem CID 123481
IUPAC Name 2-(3,5-dimethylphenyl)acetonitrile
SMILES CC1=CC(=CC(=C1)CC#N)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1017004
View Documents
Thermo Scientific Chemicals
L1017004
2 g
Each for $136.10
Only null left
 
AAL1017009
View Documents
Thermo Scientific Chemicals
L1017009
10 g
Each for $490.75
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39101-54-7
145.205
LMUKNQSVBFEUKR-UHFFFAOYSA-N
123481
CC1=CC(=CC(=C1)CC#N)C
C10H11N
MFCD00060304
3,5-dimethylphenylacetonitrile, 2-3,5-dimethylphenyl acetonitrile, 3,5-dimethylphenyl acetonitrile, benzeneacetonitrile, 3,5-dimethyl, 3,5-dimethyl-phenyl-acetonitrile, 2-3,5-dimethylphenyl ethanenitrile, acmc-20a3vm, 3.5-dimethylphenylacetonitril, 3,5-dimethylbenzeneacetonitrile, 3,5-dimethylphenylethanenitrile
2-(3,5-dimethylphenyl)acetonitrile

Specifications

39101-54-7
170°C (20 mmHg)
MFCD00060304
UN3439
3,5-dimethylphenylacetonitrile, 2-3,5-dimethylphenyl acetonitrile, 3,5-dimethylphenyl acetonitrile, benzeneacetonitrile, 3,5-dimethyl, 3,5-dimethyl-phenyl-acetonitrile, 2-3,5-dimethylphenyl ethanenitrile, acmc-20a3vm, 3.5-dimethylphenylacetonitril, 3,5-dimethylbenzeneacetonitrile, 3,5-dimethylphenylethanenitrile
CC1=CC(=CC(=C1)CC#N)C
145.205
145.2
3,5-Dimethylphenylacetonitrile
42°C to 45°C
C10H11N
2 g
2573956
LMUKNQSVBFEUKR-UHFFFAOYSA-N
2-(3,5-dimethylphenyl)acetonitrile
123481
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 254-292-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only