missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylacetonitrile, 98%

Catalog No. AAL1017004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1017004 2 g
AAL1017009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1017004 Supplier Thermo Scientific Chemicals Supplier No. L1017004

CAS: 39101-54-7 | C10H11N | 145.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39101-54-7
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00060304
InChI Key LMUKNQSVBFEUKR-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylacetonitrile, 2-3,5-dimethylphenyl acetonitrile, 3,5-dimethylphenyl acetonitrile, benzeneacetonitrile, 3,5-dimethyl, 3,5-dimethyl-phenyl-acetonitrile, 2-3,5-dimethylphenyl ethanenitrile, acmc-20a3vm, 3.5-dimethylphenylacetonitril, 3,5-dimethylbenzeneacetonitrile, 3,5-dimethylphenylethanenitrile
PubChem CID 123481
IUPAC Name 2-(3,5-dimethylphenyl)acetonitrile
SMILES CC1=CC(=CC(=C1)CC#N)C

Specifications

Melting Point 42°C to 45°C
Boiling Point 170°C (20 mmHg)
Quantity 2 g
UN Number UN3439
Beilstein 2573956
Formula Weight 145.2
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 254-292-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only