missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol 99.0+%, TCI America™

Catalog No. p-7126427
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D077825G 25 g
D0778500G 500 g
2 options

Catalog No. D077825G

Supplier: TCI America D077825G

Only null left

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 220°C
Quantity 25 g
UN Number 2261
Formula Weight 122.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE6475000
TSCA Yes