missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol, 99+%

CAS: 108-68-9 | C8H10O | 122.17 g/mol

Supplier:  Thermo Scientific Chemicals 162791000

 View more versions of this product

Catalog No. AC162791000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3, 5-Dimethylphenol
108-68-9
100.0
Orange to White
222°C
Authentic
Glass bottle
(CH3)2C6H3OH
100 g
1.115
3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
TUAMRELNJMMDMT-UHFFFAOYSA-N
3,5-dimethylphenol
7948
122.17
Crystalline Solid
99+%
99.0
63°C to 66°C
1.1150g/mL
109°C
99% min. (GC)
C8H10O
MFCD00002307
06, 492
15, 10279
Solubility in water: 5.3g/L (25°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
CC1=CC(O)=CC(C)=C1
122.17
CHEBI:38572
99+%

Chemical Identifiers

108-68-9
122.17
TUAMRELNJMMDMT-UHFFFAOYSA-N
7948
3,5-dimethylphenol
C8H10O
MFCD00002307
3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
CHEBI:38572
CC1=CC(O)=CC(C)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-606-5

RTECSNumber : ZE6475000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only