missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzyl alcohol, 99%

Catalog No. p-7046520
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1472603 1 g
AAL1472606 5 g
AAL1472614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1472603 Supplier Thermo Scientific Chemicals Supplier No. L1472603

CAS: 27129-87-9 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27129-87-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004648
InChI Key IQWWTJDRVBWBEL-UHFFFAOYSA-N
PubChem CID 33706
IUPAC Name (3,5-dimethylphenyl)methanol
SMILES CC1=CC(=CC(=C1)CO)C

Specifications

Boiling Point 218°C to 221°C
Flash Point 106°C (222°F)
Refractive Index 1.531
Quantity 1 g
Beilstein 2077285
Synonym 3,5-dimethylphenyl methanol, 3,5-dimethylbenzyl alcohol, 3,5-dimethylbenzenemethanol, 3,5-dimethylbenzylalcohol, benzenemethanol, 3,5-dimethyl, benzyl alcohol, 3,5-dimethyl, 3,5-dimethylphenyl methan-1-ol, acmc-1cr8g, 3,5-dimethyl benzyl alcohol, benzenemethanol,3,5-dimethyl
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylbenzyl alcohol
EINECSNumber 248-241-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only