accessibility menu, dialog, popup

UserName

CH3CH3OH

CAS RN 27129-87-9

IUPAC Name: (3,5-dimethylphenyl)methanol
Synonyms: benzenemethanol, 3,5-dimethyl-
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: IQWWTJDRVBWBEL-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=CC(C)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results