missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzeneboronic acid, 98%

CAS: 172975-69-8 | C8H11BO2 | 149.984 g/mol

$81.73 - $661.99

Chemical Identifiers

CAS 172975-69-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD00185689
InChI Key DJGHSJBYKIQHIK-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
PubChem CID 2734349
IUPAC Name (3,5-dimethylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C)C)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2343403
View Documents
Thermo Scientific Chemicals
B2343403
1 g
Each for $81.73
Only null left
 
AAB2343406
View Documents
Thermo Scientific Chemicals
B2343406
5 g
Each for $197.71
Only null left
 
AAB2343414
View Documents
Thermo Scientific Chemicals
B2343414
25 g
Each for $661.99
Only null left
 
Description

Description

3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Chemical Identifiers

172975-69-8
149.984
DJGHSJBYKIQHIK-UHFFFAOYSA-N
2734349
B(C1=CC(=CC(=C1)C)C)(O)O
C8H11BO2
MFCD00185689
3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
(3,5-dimethylphenyl)boronic acid

Specifications

235°C to 237°C
1 g
MFCD00185689
3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
B(C1=CC(=CC(=C1)C)C)(O)O
149.984
149.99
3,5-Dimethylbenzeneboronic acid
172975-69-8
C8H11BO2
7368658
DJGHSJBYKIQHIK-UHFFFAOYSA-N
(3,5-dimethylphenyl)boronic acid
2734349
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only