missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzaldehyde 96.0+%, TCI America™

Catalog No. p-7127761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34985G 5 g
D349825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34985G Supplier TCI America Supplier No. D34985G
Only null left

Chemical Identifiers

CAS 5779-95-3
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00082777
InChI Key NBEFMISJJNGCIZ-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
PubChem CID 34225
IUPAC Name 3,5-dimethylbenzaldehyde
SMILES CC1=CC(=CC(=C1)C=O)C

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylbenzaldehyde