accessibility menu, dialog, popup

UserName

CAS RN 5779-95-3

IUPAC Name: 3,5-dimethylbenzaldehyde
Synonyms: 3,5-dimethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: NBEFMISJJNGCIZ-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)