missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzaldehyde, 98%

Catalog No. p-7045191
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0974803 1 g
AAL0974806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0974803 Supplier Thermo Scientific Chemicals Supplier No. L0974803

CAS: 5779-95-3 | C9H10O | 134.178 g/mol

3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

Solubility
Not miscible with water.

Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.

Chemical Identifiers

CAS 5779-95-3
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00082777
InChI Key NBEFMISJJNGCIZ-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
PubChem CID 34225
IUPAC Name 3,5-dimethylbenzaldehyde
SMILES CC1=CC(=CC(=C1)C=O)C

Specifications

Melting Point 8°C to 10°C
Boiling Point 220°C to 222°C
Refractive Index 1.5385
Quantity 1 g
Beilstein 2038840
Sensitivity Air Sensitive
Solubility Information Not miscible with water.
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylbenzaldehyde
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 675-424-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only