Learn More
3,5-Dimethylbenzaldehyde, 98%
CAS: 5779-95-3 | C9H10O | 134.178 g/mol
Supplier: Thermo Scientific Chemicals L0974806
Description
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.
Solubility
Not miscible with water.
Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.
Specifications
| 3,5-Dimethylbenzaldehyde | |
| 8°C to 10°C | |
| C9H10O | |
| MFCD00082777 | |
| 2038840 | |
| benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde | |
| NBEFMISJJNGCIZ-UHFFFAOYSA-N | |
| 3,5-dimethylbenzaldehyde | |
| 34225 | |
| ≥98% |
| 5779-95-3 | |
| 220°C to 222°C | |
| 1.5385 | |
| 5 g | |
| Air Sensitive | |
| Not miscible with water. | |
| CC1=CC(=CC(=C1)C=O)C | |
| 134.178 | |
| 134.18 |
Chemical Identifiers
| 5779-95-3 | |
| 134.178 | |
| NBEFMISJJNGCIZ-UHFFFAOYSA-N | |
| 34225 | |
| CC1=CC(=CC(=C1)C=O)C |
| C9H10O | |
| MFCD00082777 | |
| benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde | |
| 3,5-dimethylbenzaldehyde |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 675-424-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only