missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxyphenylacetonitrile 98.0+%, TCI America™
Supplier: TCI America D51865G
Specifications
| 3,5-Dimethoxyphenylacetonitrile | |
| 56°C | |
| 178°C | |
| MFCD00016395 | |
| 3,5-dimethoxyphenylacetonitrile, 2-3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxy-benzeneacetonitrile, benzeneacetonitrile, 3,5-dimethoxy, 2-3,5-dimethoxyphenyl ethanenitrile, pubchem16207, 3,5-dimethoxybenzylcyanide, acmc-20a41u | |
| COC1=CC(OC)=CC(CC#N)=C1 | |
| 177.20 | |
| 177.20 | |
| Crystalline Powder |
| 13388-75-5 | |
| White-Yellow | |
| C10H11NO2 | |
| 5 g | |
| UUNRWZQWCNTSCV-UHFFFAOYSA-N | |
| 2-(3,5-dimethoxyphenyl)acetonitrile | |
| 139445 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13388-75-5 | |
| 177.20 | |
| UUNRWZQWCNTSCV-UHFFFAOYSA-N | |
| 139445 | |
| COC1=CC(OC)=CC(CC#N)=C1 |
| C10H11NO2 | |
| MFCD00016395 | |
| 3,5-dimethoxyphenylacetonitrile, 2-3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxy-benzeneacetonitrile, benzeneacetonitrile, 3,5-dimethoxy, 2-3,5-dimethoxyphenyl ethanenitrile, pubchem16207, 3,5-dimethoxybenzylcyanide, acmc-20a41u | |
| 2-(3,5-dimethoxyphenyl)acetonitrile |
Safety and Handling
TSCA : No