accessibility menu, dialog, popup

UserName

H3CONOH3C

CAS RN 13388-75-5

IUPAC Name: 2-(3,5-dimethoxyphenyl)acetonitrile
Synonyms: 2-(3,5-dimethoxyphenyl)acetonitrile
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: UUNRWZQWCNTSCV-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(CC#N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)