missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylacetonitrile 98.0+%, TCI America™

Catalog No. D51861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51861G 1 g
D51865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51861G Supplier TCI America Supplier No. D51861G

Chemical Identifiers

CAS 13388-75-5
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00016395
InChI Key UUNRWZQWCNTSCV-UHFFFAOYSA-N
Synonym 3,5-dimethoxyphenylacetonitrile, 2-3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxy-benzeneacetonitrile, benzeneacetonitrile, 3,5-dimethoxy, 2-3,5-dimethoxyphenyl ethanenitrile, pubchem16207, 3,5-dimethoxybenzylcyanide, acmc-20a41u
PubChem CID 139445
IUPAC Name 2-(3,5-dimethoxyphenyl)acetonitrile
SMILES COC1=CC(OC)=CC(CC#N)=C1

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 178°C
Quantity 1 g
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxyphenylacetonitrile