Learn More
3,5-Dimethoxybenzylamine, 98%
CAS: 34967-24-3 | C9H13NO2 | 167.208 g/mol
Supplier: Thermo Scientific Chemicals H6050314
Description
3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications
| 3,5-Dimethoxybenzylamine | |
| 35°C to 37°C | |
| 94°C to 96°C | |
| C9H13NO2 | |
| MFCD00052813 | |
| UN3259 | |
| Slightly soluble in water. | |
| COC1=CC(=CC(=C1)CN)OC | |
| 167.208 | |
| 167.21 |
| 34967-24-3 | |
| 1.106 | |
| 113°C (235°F) | |
| 1.542 | |
| 25 g | |
| 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine | |
| YGZJTYCCONJJGZ-UHFFFAOYSA-N | |
| (3,5-dimethoxyphenyl)methanamine | |
| 420973 | |
| 98% |
Chemical Identifiers
| 34967-24-3 | |
| 167.208 | |
| YGZJTYCCONJJGZ-UHFFFAOYSA-N | |
| 420973 | |
| COC1=CC(=CC(=C1)CN)OC |
| C9H13NO2 | |
| MFCD00052813 | |
| 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine | |
| (3,5-dimethoxyphenyl)methanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only