accessibility menu, dialog, popup

UserName

CAS RN 34967-24-3

IUPAC Name: 1-(3,5-dimethoxyphenyl)methanamine
Synonyms: 1-(3,5-dimethoxyphenyl)methanamine
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: YGZJTYCCONJJGZ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(CN)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)