Learn More
3,5-Dimethoxy-4-methylbenzoic acid, 97%
CAS: 61040-81-1 | C10H12O4 | 196.202 g/mol
Supplier: Thermo Scientific Chemicals A1480114
Description
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.
Solubility
Soluble in liquid ammonia.
Notes
Incompatible with oxidizing agents.
Specifications
| 3,5-Dimethoxy-4-methylbenzoic acid | |
| 210°C to 216°C | |
| MFCD00017506 | |
| 2695238 | |
| Soluble in liquid ammonia. | |
| CC1=C(C=C(C=C1OC)C(=O)O)OC | |
| 196.202 | |
| 196.2 |
| 61040-81-1 | |
| C10H12O4 | |
| 25 g | |
| 3,5-dimethoxy-4-methyl-benzoic acid, benzoic acid,3,5-dimethoxy-4-methyl, 3,5-dimethoxy-4-methylbenzoicacid, acmc-1b58y, 3,5-dimethoxy4-methylbenzoic acid, 4-methyl-3,5-dimethoxybenzoic acid, 3,5-dimethoxy-p-toluic acid, labotest-bb lt00455447, 3,5-dimethoxy-4-methyl benzoic acid, 3,5-di-methoxy-4-methyl-benzoic acid | |
| QIBMVRYNEXOCCF-UHFFFAOYSA-N | |
| 3,5-dimethoxy-4-methylbenzoic acid | |
| 3764486 | |
| 97% |
Chemical Identifiers
| 61040-81-1 | |
| 196.202 | |
| QIBMVRYNEXOCCF-UHFFFAOYSA-N | |
| 3764486 | |
| CC1=C(C=C(C=C1OC)C(=O)O)OC |
| C10H12O4 | |
| MFCD00017506 | |
| 3,5-dimethoxy-4-methyl-benzoic acid, benzoic acid,3,5-dimethoxy-4-methyl, 3,5-dimethoxy-4-methylbenzoicacid, acmc-1b58y, 3,5-dimethoxy4-methylbenzoic acid, 4-methyl-3,5-dimethoxybenzoic acid, 3,5-dimethoxy-p-toluic acid, labotest-bb lt00455447, 3,5-dimethoxy-4-methyl benzoic acid, 3,5-di-methoxy-4-methyl-benzoic acid | |
| 3,5-dimethoxy-4-methylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only