missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-methylbenzoic acid, 97%

Catalog No. p-7022167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1480106 5 g
AAA1480114 25 g
AAA1480122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1480106 Supplier Thermo Scientific Chemicals Supplier No. A1480106

CAS: 61040-81-1 | C10H12O4 | 196.202 g/mol

3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

Solubility
Soluble in liquid ammonia.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 61040-81-1
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00017506
InChI Key QIBMVRYNEXOCCF-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-4-methyl-benzoic acid, benzoic acid,3,5-dimethoxy-4-methyl, 3,5-dimethoxy-4-methylbenzoicacid, acmc-1b58y, 3,5-dimethoxy4-methylbenzoic acid, 4-methyl-3,5-dimethoxybenzoic acid, 3,5-dimethoxy-p-toluic acid, labotest-bb lt00455447, 3,5-dimethoxy-4-methyl benzoic acid, 3,5-di-methoxy-4-methyl-benzoic acid
PubChem CID 3764486
IUPAC Name 3,5-dimethoxy-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1OC)C(=O)O)OC

Specifications

Melting Point 210°C to 216°C
Quantity 5 g
Beilstein 2695238
Solubility Information Soluble in liquid ammonia.
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material 3,5-Dimethoxy-4-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only