missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxytoluene, 99%

Catalog No. p-7047299
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1856706 5 g
AAL1856714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1856706 Supplier Thermo Scientific Chemicals Supplier No. L1856706
Only null left

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol

Used in production of orcein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-15-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002291
InChI Key OIPPWFOQEKKFEE-UHFFFAOYSA-N
Synonym orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
PubChem CID 10436
ChEBI CHEBI:16536
IUPAC Name 5-methylbenzene-1,3-diol
SMILES CC1=CC(O)=CC(O)=C1

Specifications

Melting Point 106°C to 108°C
Boiling Point 287°C to 290°C
Quantity 5 g
Beilstein 1071903
Sensitivity Air Sensitive
Merck Index 14,6864
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 3,5-Dihydroxytoluene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-984-2
RTECSNumber VH2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only