missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzamide 98.0+%, TCI America™

Catalog No. D28755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28755G Supplier TCI America Supplier No. D28755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3147-62-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017123
InChI Key PLYYMFBDRBSPJZ-UHFFFAOYSA-N
Synonym benzamide, 3,5-dihydroxy, 3,5-dihydroxy-benzamide, alpha-resorcylamide, 3,5-resorcylic acidamide, 3,5-dihydroxybenzoic acid amide, .alpha.-resorcylamide, pubchem13928, a-resorcyl amide, acmc-1coua, ksc494k8l
PubChem CID 76604
IUPAC Name 3,5-dihydroxybenzamide
SMILES C1=C(C=C(C=C1O)O)C(=O)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dihydroxybenzamide