CAS RN 3147-62-4
IUPAC Name:
3,5-dihydroxybenzamide
Synonyms:
3,5-dihydroxybenzamide
Molecular Weight (g/mol):
153.14
Molecular Formula:
C7H7NO3
InChi Key:
PLYYMFBDRBSPJZ-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC(O)=CC(O)=C1
1
–
1
of
1
results
1
3,5-Dihydroxybenzamide 98.0+%, TCI America™
CAS: 3147-62-4 Molecular Formula: C7H7NO3 Molecular Weight (g/mol): 153.137 MDL Number: MFCD00017123 InChI Key: PLYYMFBDRBSPJZ-UHFFFAOYSA-N Synonym: benzamide, 3,5-dihydroxy,3,5-dihydroxy-benzamide,alpha-resorcylamide,3,5-resorcylic acidamide,3,5-dihydroxybenzoic acid amide,.alpha.-resorcylamide,pubchem13928,a-resorcyl amide,acmc-1coua,ksc494k8l PubChem CID: 76604 IUPAC Name: 3,5-dihydroxybenzamide SMILES: C1=C(C=C(C=C1O)O)C(=O)N