missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorophenol 98.0+%, TCI America™
$93.21 - $281.48
Chemical Identifiers
| CAS | 2713-34-0 |
|---|---|
| Molecular Formula | C6H4F2O |
| Molecular Weight (g/mol) | 130.09 |
| MDL Number | MFCD00002255 |
| InChI Key | HJSSBIMVTMYKPD-UHFFFAOYSA-N |
| Synonym | phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc |
| PubChem CID | 75928 |
| IUPAC Name | 3,5-difluorophenol |
| SMILES | OC1=CC(F)=CC(F)=C1 |
Chemical Identifiers
| 2713-34-0 | |
| 130.09 | |
| HJSSBIMVTMYKPD-UHFFFAOYSA-N | |
| 75928 | |
| OC1=CC(F)=CC(F)=C1 |
| C6H4F2O | |
| MFCD00002255 | |
| phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc | |
| 3,5-difluorophenol |
Specifications
| 2713-34-0 | |
| 68°C | |
| MFCD00002255 | |
| phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc | |
| OC1=CC(F)=CC(F)=C1 | |
| 130.09 | |
| 130.09 | |
| Crystalline Powder |
| 54°C | |
| C6H4F2O | |
| 5 g | |
| HJSSBIMVTMYKPD-UHFFFAOYSA-N | |
| 3,5-difluorophenol | |
| 75928 | |
| ≥98.0% (GC) | |
| 3,5-Difluorophenol |
Safety and Handling
TSCA : No