missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol, 98+%

Catalog No. AAA1517703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1517703 1 g
AAA1517706 5 g
AAA1517714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1517703 Supplier Thermo Scientific Chemicals Supplier No. A1517703

CAS: 2713-34-0 | C6H4F2O | 130.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2713-34-0
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00002255
InChI Key HJSSBIMVTMYKPD-UHFFFAOYSA-N
PubChem CID 75928
IUPAC Name 3,5-difluorophenol
SMILES OC1=CC(F)=CC(F)=C1

Specifications

Melting Point 51°C to 55°C
Boiling Point 65°C to 68°C (1 mmHg)
Flash Point 70°C (158°F)
Odor Phenol-like
Quantity 1 g
UN Number UN3261
Beilstein 2078616
Synonym phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorophenol
Show More Show Less
Hazard Category H228-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only