missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoronitrobenzene 98.0+%, TCI America™

Catalog No. p-7127727
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34265G 5 g
D342625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34265G Supplier TCI America Supplier No. D34265G

Chemical Identifiers

CAS 2265-94-3
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00012142
InChI Key AUQBBDWDLJSKMI-UHFFFAOYSA-N
Synonym 3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
PubChem CID 75272
IUPAC Name 1,3-difluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 g
Formula Weight 159.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Difluoronitrobenzene