accessibility menu, dialog, popup

UserName

CAS RN 2265-94-3

IUPAC Name: 1,3-difluoro-5-nitrobenzene
Synonyms: benzene, 1,3-difluoro-5-nitro-
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: AUQBBDWDLJSKMI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results