missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorobenzoyl Chloride 98.0+%, TCI America™
$73.16 - $215.62
Chemical Identifiers
| CAS | 129714-97-2 |
|---|---|
| Molecular Formula | C7H3ClF2O |
| Molecular Weight (g/mol) | 176.547 |
| MDL Number | MFCD00010309 |
| InChI Key | OYZWEOORLJBPMA-UHFFFAOYSA-N |
| Synonym | 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride |
| PubChem CID | 145600 |
| IUPAC Name | 3,5-difluorobenzoyl chloride |
| SMILES | C1=C(C=C(C=C1F)F)C(=O)Cl |
Chemical Identifiers
| 129714-97-2 | |
| 176.547 | |
| OYZWEOORLJBPMA-UHFFFAOYSA-N | |
| 145600 | |
| C1=C(C=C(C=C1F)F)C(=O)Cl |
| C7H3ClF2O | |
| MFCD00010309 | |
| 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride | |
| 3,5-difluorobenzoyl chloride |
Specifications
| 129714-97-2 | |
| C7H3ClF2O | |
| 5 g | |
| 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride | |
| C1=C(C=C(C=C1F)F)C(=O)Cl | |
| 176.547 | |
| 176.55 | |
| Liquid |
| Yellow | |
| MFCD00010309 | |
| 3265 | |
| OYZWEOORLJBPMA-UHFFFAOYSA-N | |
| 3,5-difluorobenzoyl chloride | |
| 145600 | |
| ≥98.0% (GC,T) | |
| 3,5-Difluorobenzoyl Chloride |
Safety and Handling
TSCA : No