accessibility menu, dialog, popup

UserName

CAS RN 129714-97-2

IUPAC Name: 3,5-difluorobenzoyl chloride
Synonyms: 3,5-difluorobenzoyl chloride
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: OYZWEOORLJBPMA-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1)C(Cl)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)