missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D34075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34075G 5 g
D340725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34075G Supplier TCI America Supplier No. D34075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129714-97-2
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00010309
InChI Key OYZWEOORLJBPMA-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
PubChem CID 145600
IUPAC Name 3,5-difluorobenzoyl chloride
SMILES C1=C(C=C(C=C1F)F)C(=O)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 176.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzoyl Chloride