missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7127390
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25965G 5 g
D259625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25965G Supplier TCI America Supplier No. D25965G
Only null left

Chemical Identifiers

CAS 455-40-3
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00010323
InChI Key GONAVIHGXFBTOZ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoicacid, benzoic acid, 3,5-difluoro, pubchem2173, 3,5 difluorobenzoic acid, 3,5-difluoro benzoicacid, acmc-209k3a, 3,5 difluoro benzoic acid, 3,5-di-fluorobenzoic acid, 3,5-difluoro-benzoic acid
PubChem CID 101265
IUPAC Name 3,5-difluorobenzoic acid
SMILES C1=C(C=C(C=C1F)F)C(=O)O

Specifications

Melting Point 122°C
Color Purple-White
Quantity 5 g
Formula Weight 158.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorobenzoic Acid