accessibility menu, dialog, popup

UserName

CAS RN 455-40-3

IUPAC Name: 3,5-difluorobenzoic acid
Synonyms: 3,5-difluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: GONAVIHGXFBTOZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results