missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America D262625G
Specifications
| 3,5-Difluorobenzaldehyde | |
| White | |
| MFCD00010329 | |
| 1325 | |
| ASOFZHSTJHGQDT-UHFFFAOYSA-N | |
| 3,5-difluorobenzaldehyde | |
| 588160 | |
| ≥98.0% (GC) |
| 32085-88-4 | |
| C7H4F2O | |
| 25 g | |
| benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone | |
| C1=C(C=C(C=C1F)F)C=O | |
| 142.105 | |
| 142.10 | |
| Crystalline Lumps |
Chemical Identifiers
| 32085-88-4 | |
| 142.105 | |
| ASOFZHSTJHGQDT-UHFFFAOYSA-N | |
| 588160 | |
| C1=C(C=C(C=C1F)F)C=O |
| C7H4F2O | |
| MFCD00010329 | |
| benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone | |
| 3,5-difluorobenzaldehyde |
Safety and Handling
TSCA : No