missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7127409
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26265G 5 g
D262625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26265G Supplier TCI America Supplier No. D26265G
Only null left

Chemical Identifiers

CAS 32085-88-4
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010329
InChI Key ASOFZHSTJHGQDT-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone
PubChem CID 588160
IUPAC Name 3,5-difluorobenzaldehyde
SMILES C1=C(C=C(C=C1F)F)C=O

Specifications

Color White
Quantity 5 g
UN Number 1325
Formula Weight 142.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Difluorobenzaldehyde