missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-methoxyaniline, 97%

Catalog No. AAH2677806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2677806 5 g
AAH2677803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2677806 Supplier Thermo Scientific Chemicals Supplier No. H2677806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 363-47-3 | C7H7F2NO | 159.14 g/mol

3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 363-47-3
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD04115910
InChI Key POVSDXPEJZMSEJ-UHFFFAOYSA-N
Synonym 4-amino-2,6-fluoroanisole, benzenamine, 3,5-difluoro-4-methoxy-9ci, 3,5-difluoro-4-methoxy-aniline, benzenamine,3,5-difluoro-4-methoxy, pubchem10104, 3,5-difluoro-4-methxoyaniline, 3,5-difluoro-4-methoxybenzenamine, 3,5-difluoro-4-methoxyphenylamine, 3,5-difluoro-4-methoxybenzeneamine, 3,5-difluoro-4-methoxy-benzenamine
PubChem CID 2783137
IUPAC Name 3,5-difluoro-4-methoxyaniline
SMILES COC1=C(F)C=C(N)C=C1F

Specifications

Melting Point 77°C to 80°C
Odor Characteristic
Quantity 5 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxyaniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only