CAS RN 363-47-3
IUPAC Name:
3,5-difluoro-4-methoxyaniline
Synonyms:
3,5-difluoro-4-methoxyaniline
Molecular Weight (g/mol):
159.14
Molecular Formula:
C7H7F2NO
InChi Key:
POVSDXPEJZMSEJ-UHFFFAOYSA-N
SMILES:
COC1=C(F)C=C(N)C=C1F
1
–
1
of
1
results
1
Promotions Available
3,5-Difluoro-4-methoxyaniline, 97%
3,5-Difluoro-4-methoxyaniline, 97%
CAS: 363-47-3 Molecular Formula: C7H7F2NO Molecular Weight (g/mol): 159.14 MDL Number: MFCD04115910 InChI Key: POVSDXPEJZMSEJ-UHFFFAOYSA-N Synonym: 4-amino-2,6-fluoroanisole,benzenamine, 3,5-difluoro-4-methoxy-9ci,3,5-difluoro-4-methoxy-aniline,benzenamine,3,5-difluoro-4-methoxy,pubchem10104,3,5-difluoro-4-methxoyaniline,3,5-difluoro-4-methoxybenzenamine,3,5-difluoro-4-methoxyphenylamine,3,5-difluoro-4-methoxybenzeneamine,3,5-difluoro-4-methoxy-benzenamine PubChem CID: 2783137 IUPAC Name: 3,5-difluoro-4-methoxyaniline SMILES: COC1=C(F)C=C(N)C=C1F