Learn More
3,5-Difluoro-4-hydroxybenzaldehyde, 97%
CAS: 118276-06-5 | C7H4F2O2 | 158.104 g/mol
Supplier: Thermo Scientific Chemicals H2613903
Description
3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Specifications
| 3,5-Difluoro-4-hydroxybenzaldehyde | |
| 121°C to 124°C | |
| MFCD06797917 | |
| Air Sensitive | |
| SKOYTQILPMNZQO-UHFFFAOYSA-N | |
| 3,5-difluoro-4-hydroxybenzaldehyde | |
| 10192584 | |
| 99% |
| 118276-06-5 | |
| C7H4F2O2 | |
| 1 g | |
| 3,5-difluoro-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-difluorobenzaldehyde, benzaldehyde, 3,5-difluoro-4-hydroxy, pubchem17015, acmc-1c2v9, ksc495m7b, benzaldehyde, 3,5-difluoro-4-hydroxy-9ci, s-3-hydroxy-1-hydroxymethylpropyl carbamic acid benzyl ester | |
| C1=C(C=C(C(=C1F)O)F)C=O | |
| 158.104 | |
| 158.11 |
Chemical Identifiers
| 118276-06-5 | |
| 158.104 | |
| SKOYTQILPMNZQO-UHFFFAOYSA-N | |
| 10192584 | |
| C1=C(C=C(C(=C1F)O)F)C=O |
| C7H4F2O2 | |
| MFCD06797917 | |
| 3,5-difluoro-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-difluorobenzaldehyde, benzaldehyde, 3,5-difluoro-4-hydroxy, pubchem17015, acmc-1c2v9, ksc495m7b, benzaldehyde, 3,5-difluoro-4-hydroxy-9ci, s-3-hydroxy-1-hydroxymethylpropyl carbamic acid benzyl ester | |
| 3,5-difluoro-4-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only