CAS RN 118276-06-5
IUPAC Name:
3,5-difluoro-4-hydroxybenzaldehyde
Synonyms:
3,5-difluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol):
158.1
Molecular Formula:
C7H4F2O2
InChi Key:
SKOYTQILPMNZQO-UHFFFAOYSA-N
SMILES:
OC1=C(F)C=C(C=O)C=C1F
1
–
1
of
1
results
1
3,5-Difluoro-4-hydroxybenzaldehyde, 97%
CAS: 118276-06-5 Molecular Formula: C7H4F2O2 Molecular Weight (g/mol): 158.104 MDL Number: MFCD06797917 InChI Key: SKOYTQILPMNZQO-UHFFFAOYSA-N Synonym: 3,5-difluoro-4-hydroxy-benzaldehyde,4-hydroxy-3,5-difluorobenzaldehyde,benzaldehyde, 3,5-difluoro-4-hydroxy,pubchem17015,acmc-1c2v9,ksc495m7b,benzaldehyde, 3,5-difluoro-4-hydroxy-9ci,s-3-hydroxy-1-hydroxymethylpropyl carbamic acid benzyl ester PubChem CID: 10192584 IUPAC Name: 3,5-difluoro-4-hydroxybenzaldehyde SMILES: C1=C(C=C(C(=C1F)O)F)C=O