missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-hydroxybenzaldehyde, 97%

Catalog No. p-7028733
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26139MD 250 mg
AAH2613903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26139MD Supplier Thermo Scientific Chemicals Supplier No. H26139MD

CAS: 118276-06-5 | C7H4F2O2 | 158.104 g/mol

3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acid and bases.

Chemical Identifiers

CAS 118276-06-5
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD06797917
InChI Key SKOYTQILPMNZQO-UHFFFAOYSA-N
Synonym 3,5-difluoro-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-difluorobenzaldehyde, benzaldehyde, 3,5-difluoro-4-hydroxy, pubchem17015, acmc-1c2v9, ksc495m7b, benzaldehyde, 3,5-difluoro-4-hydroxy-9ci, s-3-hydroxy-1-hydroxymethylpropyl carbamic acid benzyl ester
PubChem CID 10192584
IUPAC Name 3,5-difluoro-4-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1F)O)F)C=O

Specifications

Melting Point 121°C to 124°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 158.11
Percent Purity 99%
Chemical Name or Material 3,5-Difluoro-4-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only