missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorosalicylic Acid 98.0+%, TCI America™

Catalog No. D1112500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1112500G 500 g
D111225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1112500G Supplier TCI America Supplier No. D1112500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 320-72-9
Molecular Formula C7H4Cl2O3
Molecular Weight (g/mol) 207.006
MDL Number MFCD00002442
InChI Key CNJGWCQEGROXEE-UHFFFAOYSA-N
Synonym 3,5-dichlorosalicylic acid, benzoic acid, 3,5-dichloro-2-hydroxy, salicylic acid, 3,5-dichloro, usaf do-68, 2-hydroxy-3,5-dichlorobenzoic acid, unii-o6pxr32g3v, 3,5-dichlorosalicyclic acid, 3,5-dichlorosalicylicacid, 3,5-dichloro-2-hydroxy-benzoic acid, o6pxr32g3v
PubChem CID 9445
IUPAC Name 3,5-dichloro-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Cl)O)C(=O)O)Cl

Specifications

Melting Point 227°C
Color White
Quantity 500 g
Formula Weight 207.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorosalicylic Acid
RTECSNumber VO2450000
TSCA Yes