accessibility menu, dialog, popup

UserName

CAS RN 320-72-9

IUPAC Name: 3,5-dichloro-2-hydroxybenzoic acid
Synonyms: 3,5-dichloro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 207.01
Molecular Formula: C7H4Cl2O3
InChi Key: CNJGWCQEGROXEE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC(Cl)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results